missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodate Volumetric Solution, 0.0500 M (M/20), 0.300 N, Ricca Chemical

Catalog No. R62770500C
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R62770500C 500 mL
627732 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R62770500C Supplier Ricca Chemical Company Supplier No. 627716

Chemical Identifiers

CAS 7758-05-6 , 7732-18-5
Molecular Formula IKO3
Molecular Weight (g/mol) 214.00
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
Synonym potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
PubChem CID 23665710

Specifications

CAS Min % 97.0, 1.09
CAS Max % 100.0, 1.13
SMILES [O-]I(=O)=O.[K+]
IUPAC Name potassium;iodate
Grade USP
Packaging Amber Glass Bottle
Quantity 500 mL
Chemical Name or Material POTASSIUM IODATE VOLUMETRIC SOLUTION