missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodate-Iodide, 0.0125 N (N/80), 1 mL = 0.5 mg SO32-, Ricca Chemical

Catalog No. 628016
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
628016 500 mL Amber Glass Bottle
628032 1 L Amber Glass Bottle
62801 4 L Amber Glass Bottle
R62800004F 4 L Cubitainer
62805 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 628016 Supplier Ricca Chemical Company Supplier No. 628016

Specifications

Chemical Name or Material Potassium Iodate-Iodide
Color Colorless
CAS 7732-18-5, 7681-11-0, 7758-05-6, 1310-73-2
CAS Min % 98.0, 0.42, 0.05, 0.0016
CAS Max % 100.0, 0.44, 0.05, 0.0016
Synonym dihydrogen oxide, dihydrogen monoxide
Molecular Weight (g/mol) 214 g/mol
Equivalence 1 mL = 0.5 mg SO
Grade Laboratory
Packaging Amber Glass Bottle
pH Slightly Alkaline
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 0.0125Normal
Show More Show Less