missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium iodate, 0.1N Standardized Solution
CAS: 7758-05-6 | IKO3 | 214.00 g/mol
$130.58 - $289.00
Chemical Identifiers
| Molecular Formula | IKO3 |
|---|---|
| Molecular Weight (g/mol) | 214.00 |
| MDL Number | MFCD00011406 |
| InChI Key | JLKDVMWYMMLWTI-UHFFFAOYSA-M |
| Synonym | potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat |
| PubChem CID | 23665710 |
| IUPAC Name | potassium;iodate |
| SMILES | [O-]I(=O)=O.[K+] |
Description
Potassium iodate 0.1N solution is used as a standard solution in volumetric titrations. It is extensively used in analytical chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| IKO3 | |
| MFCD00011406 | |
| potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat | |
| potassium;iodate |
| 214.00 | |
| JLKDVMWYMMLWTI-UHFFFAOYSA-M | |
| 23665710 | |
| [O-]I(=O)=O.[K+] |
Specifications
| Colorless | |
| IKO3 | |
| 0.1N | |
| Miscible with water,potassium iodide solution and sulfuric acid. | |
| [O-]I(=O)=O.[K+] | |
| 214.00 | |
| 214 | |
| 1 L | |
| Potassium iodate, Standardized Solution |
| 7758-05-6 , 7732-18-5 | |
| MFCD00011406 | |
| potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat | |
| JLKDVMWYMMLWTI-UHFFFAOYSA-M | |
| potassium;iodate | |
| 23665710 | |
| Odorless | |
| Liquid |
Safety and Handling
EINECSNumber : 231-831-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only