missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxyde de potassium, dans le méthanol, certifié, 0,500N ±0,010N (0,5M), LabChem™
Spécifications
Spécifications
| Nom ou substance chimique | Potassium Hydroxide |
| Nom Remarque | dans le méthanol |
| Couleur | Incolore |
| CAS % maximal | 97.25, 2.75 |
| Formule linéaire | KOH |
| Concentration | 0,500N ±0,010N (0,5 M) |
| Numéro UN | UN2924 |
| Densité | 0.8g/mL |
| Renseignements sur la solubilité | Soluble dans l’eau |
| Quantité | 500 mL |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Highly flammable liquid and vapor. Toxic if swallowed, in contact with skin or if inhaled. Causes skin burns and eye damage. Causes damage to organs (central nervous system, optic nerve, liver, kidneys), (dermal, oral). |
| Precautionary Statement | GHS P Statement Keep away from heat, sparks, open flames, hot surfaces. No smoking. Keep container tightly closed. Ground/bond container and receiving equipment. Use explosion-proof electrical, ventilating, lighting equipment. Use only non-sparking tools. Take precautionary measures against static discharge. Do not breathe mist, vapors, spray. Do not eat, drink or smoke when using this product. Use only outdoors or in a well-ventilated area. Wear protective gloves, protective clothing, eye protection, face protection. Wash exposed skin thoroughly after handling. If swallowed: Rinse mounth. Do not induce vomiting. Immediately call a poison center/doctor. If on skin (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. Immediately call a poison center/doctor. Wash contaminated clothing before reuse. If inhaled: Remove person to fresh air and keep comfortable for breathing. Immediately call a poison center/doctor. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. In case of fire: Use carbon dioxide (CO2), powder, alcohol-resistant foam to extinguish. Store locked up in a cool, well-ventilated place. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Température ambiante |