missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, Flakes, Technical, 90%, Spectrum™ Chemical

Numéro de catalogue. 18612720
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
18612720 500 g Poly Bottle
18612721 2.5 kg Poly Bottle
18612722 12 kg Poly Pail
18612723 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. 18612720 Fournisseur Spectrum Chemical Mfg Cor Code fournisseur P1325500GM
Il en reste null

Identifiants chimiques

CAS 1310-58-3

Spécifications

CAS Max % 1
Quantity 500 g
Percent Purity 90%
Grade Technical
Packaging Poly Bottle
Chemical Name or Material Potassium Hydroxide

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.