missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 20% (w/v), Ricca Chemical

Catalog No. 614016
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
614016 500 mL Natural Poly Bottle
R6140000100 100 mL Natural Poly Bottle
614032 1 L Natural Poly Bottle
61401 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 614016 Supplier Ricca Chemical Company Supplier No. 614016
Only null left

Chemical Identifiers

CAS 1310-58-3 , 7732-18-5
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Color Colorless
CAS Min % 71.0, 27.38
CAS Max % 74.0, 28.49
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
Quantity 500 mL
Percent Purity 19.94 to 20.06% w/v
Grade Laboratory
Packaging Natural Poly Bottle
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide Solution
ShelfLife 36 months