missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 10.0 N Solution, Spectrum™ Chemical

Numéro de catalogue. 18612738
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
18612738 1 L Poly Bottle
18612739 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 18612738 Fournisseur Spectrum Chemical Mfg Cor Code fournisseur P7901LT
Il en reste null

Identifiants chimiques

CAS 1310-58-3 , 7732-18-5

Spécifications

Concentration 10.0 N ± 0.5 N
Quantity 1 L
Grade Chemical Solution
Packaging Poly Bottle
Chemical Name or Material potassium hydroxide solution

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.