missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide, 1.00 N in Isopropyl Alcohol, Ricca Chemical
$248.47 - $248.47
Chemical Identifiers
| CAS | 1310-58-3 |
|---|---|
| Molecular Formula | HKO |
| Molecular Weight (g/mol) | 56.11 |
| MDL Number | MFCD00003553 |
| InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
| Synonym | potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 |
| PubChem CID | 14797 |
| ChEBI | CHEBI:32035 |
| IUPAC Name | potassium hydroxide |
| SMILES | [OH-].[K+] |
Chemical Identifiers
| 1310-58-3 | |
| 56.11 | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| 14797 | |
| potassium hydroxide |
| HKO | |
| MFCD00003553 | |
| potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
| CHEBI:32035 | |
| [OH-].[K+] |
Specifications
| Colorless | |
| 7.48,90.0,0.03 | |
| HKO | |
| potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
| [OH-].[K+] | |
| 56.11 | |
| CHEBI:32035 | |
| Natural Poly Bottle | |
| >13 | |
| Liquid | |
| Potassium Hydroxide |