missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide, 1.00 N in Isopropyl Alcohol, Ricca Chemical
Supplier: Ricca Chemical Company 625832
Specifications
| Potassium Hydroxide | |
| 1310-58-3 , 67-63-0 , 107-15-3 | |
| 7.79,94.0,0.03 | |
| MFCD00003553 | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| potassium hydroxide | |
| 14797 | |
| Laboratory | |
| Isopropyl Alcohol | |
| 1 L | |
| 1.00Normal |
| Colorless | |
| 7.48,90.0,0.03 | |
| HKO | |
| potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
| [OH-].[K+] | |
| 56.11 | |
| CHEBI:32035 | |
| Natural Poly Bottle | |
| >13 | |
| Liquid |
Chemical Identifiers
| 1310-58-3 | |
| 56.11 | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| 14797 | |
| potassium hydroxide |
| HKO | |
| MFCD00003553 | |
| potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
| CHEBI:32035 | |
| [OH-].[K+] |