missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 0.500 N (N/2) in Denatured Alcohol, Ricca Chemical

$589.17 - $839.63

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
6265-32
Accessories View Documents
Ricca Chemical Company
626532
1 L Natural Poly Bottle
Each for $589.17
Only null left
 
6265-1
SDS Accessories View Documents
Ricca Chemical Company
62651
4 L Natural Poly Bottle
Each for $839.63
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1310-58-3
56.11
KWYUFKZDYYNOTN-UHFFFAOYSA-M
14797
potassium hydroxide
HKO
MFCD00003553
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
CHEBI:32035
[OH-].[K+]

Specifications

Colorless
7.76,90.0,0.03
HKO
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
[OH-].[K+]
56.11
CHEBI:32035
Natural Poly Bottle
>13
Liquid
Potassium Hydroxide
1310-58-3,64-17-5,107-15-3
8.07,94.0,0.03
MFCD00003553
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide
14797
Laboratory
Denatured Alcohol
1 L
0.500N
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'