missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 0.500 N (N/2) in Denatured Alcohol, Ricca Chemical

Catalog No. 626532
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
626532 1 L Natural Poly Bottle
62651 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 626532 Supplier Ricca Chemical Company Supplier No. 626532
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1310-58-3 , 64-17-5 , 107-15-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Color Colorless
CAS Min % 7.76, 90.0, 0.03
CAS Max % 8.07, 94.0, 0.03
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Denatured Alcohol
pH >13
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 0.500N
Chemical Name or Material Potassium Hydroxide
Show More Show Less