missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 0.500 N (N/2) in Denatured Alcohol, Ricca Chemical

Supplier:  Ricca Chemical Company 62651

 View more versions of this product

Catalog No. 6265-1


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Hydroxide
1310-58-3,64-17-5,107-15-3
8.07,94.0,0.03
MFCD00003553
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide
14797
Laboratory
Denatured Alcohol
4 L
0.500N
Colorless
7.76,90.0,0.03
HKO
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
[OH-].[K+]
56.11
CHEBI:32035
Natural Poly Bottle
>13
Liquid

Chemical Identifiers

1310-58-3
56.11
KWYUFKZDYYNOTN-UHFFFAOYSA-M
14797
potassium hydroxide
HKO
MFCD00003553
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
CHEBI:32035
[OH-].[K+]
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Accessories
Promotions

Promotions