missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 0.1 N Solution, Spectrum™ Chemical

Numéro de catalogue. 18612679
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
18612679 500 mL Poly Bottle
18612680 1 L Poly Bottle
18612681 4 L Poly Bottle
18612672 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. 18612679 Fournisseur Spectrum Chemical Mfg Cor Code fournisseur P350500ML
Il en reste null

Identifiants chimiques

CAS 1310-58-3 , 7732-18-5

Spécifications

Concentration 0.1 N ± 0.005 N
Quantity 500 mL
Grade Chemical Solution
Packaging Poly Bottle
Chemical Name or Material Potassium Hydroxide Solution

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.