missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide, 0.0500 N (N/20), Ricca Chemical
Description
- Tested in an ISO 17025 Accredited Facility
- Test Reports provided with Certified Values and Measurement Uncertainty
- Tight release specifications for low lot-to-lot variability
Specifications
Specifications
| CAS Min % | 0.32, 98% |
| CAS Max % | 0.33, 100% |
| Quantity | 500 mL |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| Chemical Name or Material | Potassium Hydroxide |