missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 0.0100 N (N/100) in Isopropyl Alcohol, Ricca Chemical

Catalog No. 625332
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
625332 1 L Natural Poly Bottle
R62530004A 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 625332 Supplier Ricca Chemical Company Supplier No. 625332
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1310-58-3 , 67-63-0 , 7732-18-5 , 107-15-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Color Colorless
CAS Min % 0.08, 97.0, 1.22, 0.03
CAS Max % 0.08, 100.0, 1.27, 0.03
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Isopropyl Alcohol
pH 12
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 0.0100Normal
Chemical Name or Material Potassium Hydroxide
Show More Show Less