missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydrogen Phthalate, 10 mg/L, Reagents

Catalog No. 01334486
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334486 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01334486 Supplier Labchem Inc Supplier No. CP0810001C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Potassium Hydrogen Phthalate, 10 mg/L, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Potassium Acid Phthalate, KHP, Potassium Biphthalate
IUPAC Name water
SMILES O

Specifications

pH 4
Boiling Point 100°C
Color Colorless
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 1
Grade Reagent
Chemical Name or Material Potassium Hydrogen Phthalate, 10 mg/L