missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexafluorophosphate, 95% min

Catalog No. AA11531A1
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA11531A1 1 kg
AA1153130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA11531A1 Supplier Thermo Scientific Chemicals Supplier No. 011531A1

CAS: 17084-13-8 | F6KP | 184.062 g/mol

Potassium hexafluorophosphate is a soluble neutral salt used for synthesis. It is a source of the hexafluorophosphate anion, a non-coordinating anion that confers lipophilicity to its salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17084-13-8
Molecular Formula F6KP
Molecular Weight (g/mol) 184.062
MDL Number MFCD00011412
InChI Key YZDGRYDIGCWVND-UHFFFAOYSA-N
Synonym potassium hexafluorophosphate, potassium fluophosphate, potassium hexafluorophosphate v, unii-88w436pdi9, monopotassium hexafluorophosphate, potassium hexfluorophosphate, potassium hexafluorophosphate min, phosphate 1-, hexafluoro-, potassium, potassium fluorophosphate, kpf6
PubChem CID 23688904
IUPAC Name potassium;hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.[K+]

Specifications

Melting Point 575°C
Color White
pH 3.0 to 7.0
Physical Form Crystalline
Quantity 1 kg
UN Number UN3260
Solubility Information Soluble in water (83.5mg/ml at 20°C).
Formula Weight 184.07
Percent Purity ≤95%
Odor Odorless
Sensitivity Hygroscopic
Density 2.55 g/mL
Chemical Name or Material Potassium hexafluorophosphate
Show More Show Less
Hazard Category H302+H312+H332-H314
Hazard Statement GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P330+P331-P302+P352-P304+P340-P305+P351+P338-P310-P312-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 241-143-0
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only