missing translation for 'onlineSavingsMsg'
Learn More

Potassium Fluoride, 50% (w/w), pH 8.3, Ricca Chemical

Catalog No. 610532
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
610532 1 L Natural Poly Bottle
61051 4 L Natural Poly Bottle
61055 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 610532 Supplier Ricca Chemical Company Supplier No. 610532
Only null left

Chemical Identifiers

CAS 7789-23-3 , 7732-18-5
Molecular Formula FK
Molecular Weight (g/mol) 58.10
MDL Number MFCD00011398
InChI Key NROKBHXJSPEDAR-UHFFFAOYSA-M
Synonym potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
PubChem CID 522689
ChEBI CHEBI:66872
IUPAC Name potassium fluoride
SMILES [F-].[K+]

Specifications

CAS Min % 70.0, 49.0
CAS Max % 73.0, 51.0
Percent Purity 50%
Grade Laboratory
Concentration or Composition (by Analyte or Components) 50% (w/w) Aqueous Solution
Packaging Natural Poly Bottle
pH 8.3
Quantity 1 L
Chemical Name or Material Potassium Fluoride