missing translation for 'onlineSavingsMsg'
Learn More

Potassium Fluoride, 50% (w/v), Neutralized, Ricca Chemical

$396.75 - $5,161.28

Chemical Identifiers

CAS 7789-23-3
Molecular Formula FK
Molecular Weight (g/mol) 58.10
MDL Number MFCD00011398
InChI Key NROKBHXJSPEDAR-UHFFFAOYSA-M
Synonym potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
PubChem CID 522689
ChEBI CHEBI:66872
IUPAC Name potassium fluoride
SMILES [F-].[K+]
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
610116
SDS View Documents
Ricca Chemical Company
610116
500 mL Natural Poly Bottle N/A
Only null left
 
610132
View Documents
Ricca Chemical Company
610132
1 L Natural Poly Bottle N/A
Only null left
 
61011
SDS View Documents
Ricca Chemical Company
61011
4 L Natural Poly Bottle N/A
Only null left
 
61015
SDS View Documents
Ricca Chemical Company
RI61015
20 L Cubitainer N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7789-23-3
58.10
NROKBHXJSPEDAR-UHFFFAOYSA-M
522689
potassium fluoride
FK
MFCD00011398
potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
CHEBI:66872
[F-].[K+]

Specifications

Colorless to Pink
65.0,32.68
FK
potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
[F-].[K+]
58.10
CHEBI:66872
Laboratory
Neutralized
∼7
Liquid
Potassium Fluoride, Neutralized
7789-23-3,7732-18-5
68.0,34.01
MFCD00011398
NROKBHXJSPEDAR-UHFFFAOYSA-M
potassium fluoride
522689
50%
Natural Poly Bottle
Aqueous Solution
500 mL
50% (w/v)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'