missing translation for 'onlineSavingsMsg'
Learn More

Potassium Fluoride, 50% (w/v), Neutralized, Ricca Chemical

Catalog No. 61015
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
61015 20 L Cubitainer
610116 500 mL Natural Poly Bottle
610132 1 L Natural Poly Bottle
61011 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 61015 Supplier Ricca Chemical Company Supplier No. RI61015
Only null left

Chemical Identifiers

CAS 7789-23-3 , 7732-18-5
Molecular Formula FK
Molecular Weight (g/mol) 58.10
MDL Number MFCD00011398
InChI Key NROKBHXJSPEDAR-UHFFFAOYSA-M
Synonym potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
PubChem CID 522689
ChEBI CHEBI:66872
IUPAC Name potassium fluoride
SMILES [F-].[K+]

Specifications

Color Colorless to Pink
CAS Min % 65.0, 32.68
CAS Max % 68.0, 34.01
Percent Purity 50%
Grade Laboratory
Packaging Cubitainer
Treatment(s) Neutralized
Solvent or Matrix Aqueous Solution
pH ∼7
Quantity 20 L
Physical Form Liquid
Concentration or Composition Notes 50% (w/v)
Chemical Name or Material Potassium Fluoride, Neutralized
Show More Show Less