missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium disulfite, ≥98% (RT), MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 16731-55-8 |
| Physical Form | Neat |
| Linear Formula | K2S2O5 |
| MDL Number | MFCD00167605 |
| Synonym | E224; Potassium metabisulfite; Potassium pyrosulfite |
| Solubility | Soluble in water |
| Type | Analytical Standard |
| Quantity | 1 g |
| Formula Weight | 222.32 |
| Percent Purity | ≥98% (RT) |
| Show More |