missing translation for 'onlineSavingsMsg'
Learn More

Potassium Dichromate TS, 7.5% (w/v), Ricca Chemical

Catalog No. 604116
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
604116 500 mL Natural Poly Bottle
R6041000100 100 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 604116 Supplier Ricca Chemical Company Supplier No. 604116
Only null left

Chemical Identifiers

CAS 7778-50-9 , 7732-18-5
Molecular Formula Cr2K2O7
Molecular Weight (g/mol) 294.182
InChI Key KMUONIBRACKNSN-UHFFFAOYSA-N
Synonym potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
PubChem CID 24502
ChEBI CHEBI:53444
IUPAC Name dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]

Specifications

Color Orange
CAS Min % 91.0, 6.84
CAS Max % 95.0, 7.12
Percent Purity 7.5%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
pH ∼3.5 to 4
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 7.5% (w/v)
Chemical Name or Material Potassium Dichromate 7.5% (w/v) Aqueous Solution, Test Solution
Show More Show Less