CAS RN 7778-50-9
Potassium dichromate, ACS, 99.0% min
CAS: 7778-50-9 Molecular Formula: Cr2K2O7 Molecular Weight (g/mol): 294.182 MDL Number: MFCD00011367 InChI Key: KMUONIBRACKNSN-UHFFFAOYSA-N Synonym: potassium dichromate,potassium bichromate,kaliumdichromat,iopezite,dipotassium bichromate,dipotassium dichromate,potassium dichromate vi,dichromic acid dipotassium salt,dipotassium dichromium heptaoxide,bichromate of potash PubChem CID: 24502 ChEBI: CHEBI:53444 IUPAC Name: dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium SMILES: [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
Potassium Dichromate, Certified, 0.1000N ±0.0005N (0.0167M), LabChem™
CAS: 7778-50-9 Molecular Formula: Cr2K2O7 Molecular Weight (g/mol): 294.182 InChI Key: KMUONIBRACKNSN-UHFFFAOYSA-N Synonym: potassium dichromate,potassium bichromate,kaliumdichromat,iopezite,dipotassium bichromate,dipotassium dichromate,potassium dichromate vi,dichromic acid dipotassium salt,dipotassium dichromium heptaoxide,bichromate of potash PubChem CID: 24502 ChEBI: CHEBI:53444 IUPAC Name: dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium SMILES: [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
Potassium dichromate, 0.1N Standardized Solution
CAS: 7778-50-9 Molecular Formula: Cr2K2O7 Molecular Weight (g/mol): 294.182 MDL Number: MFCD00011367 InChI Key: KMUONIBRACKNSN-UHFFFAOYSA-N PubChem CID: 24502 ChEBI: CHEBI:53444 IUPAC Name: dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium SMILES: [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
Potassium dichromate, 99%
CAS: 7778-50-9 Molecular Formula: Cr2K2O7 Molecular Weight (g/mol): 294.182 MDL Number: MFCD00011367 InChI Key: KMUONIBRACKNSN-UHFFFAOYSA-N Synonym: potassium dichromate,potassium bichromate,kaliumdichromat,iopezite,dipotassium bichromate,dipotassium dichromate,potassium dichromate vi,dichromic acid dipotassium salt,dipotassium dichromium heptaoxide,bichromate of potash PubChem CID: 24502 ChEBI: CHEBI:53444 IUPAC Name: dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium SMILES: [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
Potassium dichromate, 0.025N Standardized Solution
CAS: 7778-50-9 Molecular Formula: Cr2K2O7 Molecular Weight (g/mol): 294.182 MDL Number: MFCD00011367 InChI Key: KMUONIBRACKNSN-UHFFFAOYSA-N Synonym: potassium dichromate,potassium bichromate,kaliumdichromat,iopezite,dipotassium bichromate,dipotassium dichromate,potassium dichromate vi,dichromic acid dipotassium salt,dipotassium dichromium heptaoxide,bichromate of potash PubChem CID: 24502 ChEBI: CHEBI:53444 IUPAC Name: dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium SMILES: [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
Chromium VI - WP, MilliporeSigma™ Supelco™
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.
Chromium VI -WP, MilliporeSigma™ Supelco™
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Chromium VI - WS, MilliporeSigma™ Supelco™
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.
Potassium dichromate, 0.25N Standardized Solution
CAS: 7778-50-9 Molecular Formula: Cr2K2O7 Molecular Weight (g/mol): 294.182 MDL Number: MFCD00011367 InChI Key: KMUONIBRACKNSN-UHFFFAOYSA-N Synonym: potassium dichromate,potassium bichromate,kaliumdichromat,iopezite,dipotassium bichromate,dipotassium dichromate,potassium dichromate vi,dichromic acid dipotassium salt,dipotassium dichromium heptaoxide,bichromate of potash PubChem CID: 24502 ChEBI: CHEBI:53444 IUPAC Name: dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium SMILES: [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
Potassium dichromate, 99.5%, extra pure
CAS: 7778-50-9 Molecular Formula: Cr2K2O7 Molecular Weight (g/mol): 294.19 MDL Number: MFCD00011367 InChI Key: KMUONIBRACKNSN-UHFFFAOYSA-N Synonym: potassium dichromate,potassium bichromate,kaliumdichromat,iopezite,dipotassium bichromate,dipotassium dichromate,potassium dichromate vi,dichromic acid dipotassium salt,dipotassium dichromium heptaoxide,bichromate of potash PubChem CID: 24502 ChEBI: CHEBI:53444 IUPAC Name: dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium SMILES: [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
Potassium dichromate, 99+%, ACS reagent
CAS: 7778-50-9 MDL Number: MFCD00011367 InChI Key: KMUONIBRACKNSN-UHFFFAOYSA-N Synonym: potassium dichromate,potassium bichromate,kaliumdichromat,iopezite,dipotassium bichromate,dipotassium dichromate,potassium dichromate vi,dichromic acid dipotassium salt,dipotassium dichromium heptaoxide,bichromate of potash PubChem CID: 24502 ChEBI: CHEBI:53444 IUPAC Name: dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium SMILES: [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
Potassium dichromate, 99.5%, for analysis
CAS: 7778-50-9 Molecular Formula: Cr2K2O7 Molecular Weight (g/mol): 294.19 MDL Number: MFCD00011367 InChI Key: KMUONIBRACKNSN-UHFFFAOYSA-N Synonym: potassium dichromate,potassium bichromate,kaliumdichromat,iopezite,dipotassium bichromate,dipotassium dichromate,potassium dichromate vi,dichromic acid dipotassium salt,dipotassium dichromium heptaoxide,bichromate of potash PubChem CID: 24502 ChEBI: CHEBI:53444 IUPAC Name: dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium SMILES: [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
Potassium Dichromate, ACS Reagent Grade, Ricca Chemical
CAS: 7778-50-9 Molecular Formula: Cr2K2O7 Molecular Weight (g/mol): 294.182 InChI Key: KMUONIBRACKNSN-UHFFFAOYSA-N Synonym: potassium dichromate,potassium bichromate,kaliumdichromat,iopezite,dipotassium bichromate,dipotassium dichromate,potassium dichromate vi,dichromic acid dipotassium salt,dipotassium dichromium heptaoxide,bichromate of potash PubChem CID: 24502 ChEBI: CHEBI:53444 IUPAC Name: dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium SMILES: [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
Potassium Dichromate, 0.3945 N, Ricca Chemical
CAS: 7778-50-9 Molecular Formula: Cr2K2O7 Molecular Weight (g/mol): 294.182 InChI Key: KMUONIBRACKNSN-UHFFFAOYSA-N Synonym: potassium dichromate,potassium bichromate,kaliumdichromat,iopezite,dipotassium bichromate,dipotassium dichromate,potassium dichromate vi,dichromic acid dipotassium salt,dipotassium dichromium heptaoxide,bichromate of potash PubChem CID: 24502 ChEBI: CHEBI:53444 IUPAC Name: dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium SMILES: [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
Potassium Dichromate, 0.0250 N (N/40), Ricca Chemical
Chemical Name or Material: Potassium Dichromate