missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Dichromate, For COD, Certified, 0.0250N ±0.0005N (0.0042M), LabChem™
Specifications
Specifications
| Color | Orange |
| CAS Max % | 99.88, 0.12 |
| Linear Formula | K2Cr2O7 |
| Concentration | 0.0250N ±0.0005N (0.0042M) |
| Density | 1g/mL |
| Solubility Information | Soluble in water |
| Formula Weight | 294.18 |
| Grade | Certified |
| Identification | Passes Test |
| Packaging | Poly Bottle |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause genetic defects. May cause cancer. May damage fertility or the unborn child. |
| Precautionary Statement | GHS P Statement Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. Avoid breathing mist. Wear protective gloves, protective clothing, eye protection, face protection. Wear respiratory protection. Contaminated work clothing must not be allowed out of the workplace. If exposed or concerned: Get medical advice/attention. If on skin: Wash with plenty of soap and water. Take off contaminated clothing and wash it before reuse. If skin irritation or rash occurs: Get medical advice/attention. If inhaled: Remove person to fresh air and keep comfortable for breathing. If experiencing respiratory symptoms: Call a poison center/doctor. Store locked up. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |