missing translation for 'onlineSavingsMsg'
Learn More

Potassium Dichromate, 0.200 N (N/5), Ricca Chemical

$278.33 - $278.33

Chemical Identifiers


Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6065-32
View Documents
Ricca Chemical Company
606532
1 L
Each for $278.33
Only null left
 
Description

Description

Specifications

Chemical Identifiers

0.0

Specifications

7732-18-5
294.18 g/mol
0.200N (N/5)
∼4
Potassium Dichromate
dihydrogen oxide, dihydrogen monoxide
Laboratory
Natural Poly Bottle
1 L
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'