missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Dichromate, 0.200 N (N/5), Ricca Chemical
Specifications
Specifications
| CAS | 7732-18-5 |
| Synonym | dihydrogen oxide, dihydrogen monoxide |
| Molecular Weight (g/mol) | 294.18 g/mol |
| Grade | Laboratory |
| Concentration or Composition (by Analyte or Components) | 0.200N (N/5) |
| Packaging | Natural Poly Bottle |
| pH | ∼4 |
| Quantity | 1 L |
| Chemical Name or Material | Potassium Dichromate |