Learn More
Potassium dichromate, 0.1N Standardized Solution
CAS: 7778-50-9 | Cr2K2O7 | 294.182 g/mol
Supplier: Thermo Scientific Chemicals 035618AP
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Potassium dichromate | |
| 23 hPa (17mm Hg) at 20°C | |
| Cr2K2O7 | |
| Orange | |
| [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+] | |
| 294.182 | |
| 24502 | |
| 294.18 |
| 7778-50-9 | |
| Liquid | |
| MFCD00011367 | |
| KMUONIBRACKNSN-UHFFFAOYSA-N | |
| dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium | |
| 500 mL | |
| CHEBI:53444 |
Chemical Identifiers
| 7778-50-9 | |
| 294.182 | |
| KMUONIBRACKNSN-UHFFFAOYSA-N | |
| CHEBI:53444 | |
| [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+] |
| Cr2K2O7 | |
| MFCD00011367 | |
| 24502 | |
| dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium |
Safety and Handling
P201-P202-P260-P271-P281-P304+P340-P308+P313-P501c
H317-H332-H334-H340-H350-H360FD
EINECSNumber : 231-906-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only