missing translation for 'onlineSavingsMsg'
Learn More

Potassium dichromate, 0.1N Standardized Solution

Catalog No. p-7122654
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35618AP 500 mL
AA35618K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA35618AP Supplier Thermo Scientific Chemicals Supplier No. 035618AP
Only null left

CAS: 7778-50-9 | Cr2K2O7 | 294.182 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7778-50-9
Molecular Formula Cr2K2O7
Molecular Weight (g/mol) 294.182
MDL Number MFCD00011367
InChI Key KMUONIBRACKNSN-UHFFFAOYSA-N
PubChem CID 24502
ChEBI CHEBI:53444
IUPAC Name dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]

Specifications

Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Color Orange
Quantity 500 mL
Formula Weight 294.18
Chemical Name or Material Potassium dichromate
Hazard Category H317-H332-H334-H340-H350-H360FD
Precautionary Statement P201-P202-P260-P271-P281-P304+P340-P308+P313-P501c
EINECSNumber 231-906-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only