missing translation for 'onlineSavingsMsg'
Learn More

Potassium Dichromate, 0.100 N (N/10), 0.0167 M (M/60), Ricca Chemical

Catalog No. p-3252128
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R6060000100 100 mL Natural Poly Bottle
606016 500 mL Natural Poly Bottle
606032 1 L Natural Poly Bottle
60601 4 L Natural Poly Bottle
60605 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. R6060000100 Supplier Ricca Chemical Company Supplier No. R6060000100A
Only null left

Chemical Identifiers

CAS 7778-50-9 , 7732-18-5
Molecular Formula Cr2K2O7
Molecular Weight (g/mol) 294.182
InChI Key KMUONIBRACKNSN-UHFFFAOYSA-N
Synonym potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
PubChem CID 24502
ChEBI CHEBI:53444
IUPAC Name dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]

Specifications

Color Orange
CAS Min % 98.0, 0.48
CAS Max % 100.0, 0.5
Grade Laboratory
Packaging Natural Poly Bottle
Methods ACS/EP/USP Volumetric Solution
pH ∼4
Quantity 100 mL
Physical Form Liquid
Concentration or Composition Notes 0.100N,0.0167M
Chemical Name or Material Potassium Dichromate
Show More Show Less