missing translation for 'onlineSavingsMsg'
Learn More

Potassium Dichromate, 0.0250 N (N/40), Ricca Chemical

Catalog No. R605000020F
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R605000020F 20 L Cubitainer
605016 500 mL Natural Poly Bottle
605032 1 L Natural Poly Bottle
60501 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. R605000020F Supplier Ricca Chemical Company Supplier No. R605000020F
Only null left

For ASTM/EPA Testing

Chemical Identifiers

Molecular Formula Cr2K2O7
Molecular Weight (g/mol) 294.182
InChI Key KMUONIBRACKNSN-UHFFFAOYSA-N
PubChem CID 24502
ChEBI CHEBI:53444
IUPAC Name dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]

Specifications

Color Colorless
CAS 7778-50-9, 7732-18-5
Assay Percent Range 0.02500 ± 0.00001 N at 20°C
Solubility Information Solubility in Water: Infinite
Grade Laboratory
Odor Odorless
Packaging Cubitainer
Solvent or Matrix Aqueous Solution
pH ∼4
Quantity 20 L
Physical Form Liquid
Chemical Name or Material Potassium Dichromate, 0.0250N (N/40)
Show More Show Less