missing translation for 'onlineSavingsMsg'
Learn More

Potassium Dichromate, 0.0100 N (N/100), Ricca Chemical

Catalog No. 604532
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
604532 1 L Natural Poly Bottle
604516 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 604532 Supplier Ricca Chemical Company Supplier No. 604532
Only null left

Chemical Identifiers

CAS 7778-50-9 , 7732-18-5
Molecular Formula Cr2K2O7
Molecular Weight (g/mol) 294.182
InChI Key KMUONIBRACKNSN-UHFFFAOYSA-N
Synonym potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
PubChem CID 24502
ChEBI CHEBI:53444
IUPAC Name dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]

Specifications

Color Orange
CAS Min % 98.0, 0.05
CAS Max % 100.0, 0.05
Grade Laboratory
Packaging Natural Poly Bottle
pH ∼4
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 0.0100Normal
Chemical Name or Material Potassium Dichromate