missing translation for 'onlineSavingsMsg'
Learn More

Potassium cyanate, 97%

Catalog No. AA33356A3
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA33356A3 2 kg
AA3335622 100 g
AA3335636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA33356A3 Supplier Thermo Scientific Chemicals Supplier No. 033356A3
Only null left

CAS: 590-28-3 | CKNO | 81.12 g/mol

Potassium cyanate is used in gold mining, organic synthesis and electroplating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 590-28-3
Molecular Formula CKNO
Molecular Weight (g/mol) 81.12
MDL Number MFCD00011411
InChI Key GKKCIDNWFBPDBW-UHFFFAOYSA-M
Synonym potassium cyanate, kaliumcyanat, kocn, aero cyanate, cyanic acid, potassium salt, alicyanate, bulpur, potassiumcyanate, weedanol cyanol, caswell no. 688
PubChem CID 11378442
ChEBI CHEBI:38904
SMILES [K+].[O-]C#N

Specifications

Melting Point 700°C to 900°C (decomposition)
Color White
Physical Form Crystalline
Quantity 2 kg
Merck Index 14,7625
Solubility Information Soluble in water; Slightly soluble in alcohol
IUPAC Name potassium cyanate
Formula Weight 81.12
Percent Purity 97%
Odor Odorless
Density 2.056 g/mL
Chemical Name or Material Potassium cyanate
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 209-676-3
RTECSNumber GS6825000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only