missing translation for 'onlineSavingsMsg'
Learn More

Potassium Citrate, Monohydrate, Granular, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. 02003751
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02003751 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02003751 Supplier Avantor Supplier No. 306601

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 6100-05-6
Molecular Formula C6H7K3O8
Molecular Weight (g/mol) 324.41
InChI Key PJAHUDTUZRZBKM-UHFFFAOYSA-K
Synonym potassium citrate monohydrate, tripotassium citrate monohydrate, unii-ee90oni6ff, ee90oni6ff, citric acid tripotassium salt, urocit-k, ccris 6543, tri-potassium citrate monohydrate, potassium citrate tribasic monohydrate, citric acid, tripotassium salt, monohydrate
PubChem CID 2735208
ChEBI CHEBI:64746
IUPAC Name tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+]

Specifications

Physical Form Granules
Quantity 500 g
Formula Weight 324.42
Grade BAKER ANALYZED™ Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.98kg
Chemical Name or Material Potassium Citrate, Monohydrate