missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chromate TS, 10% (w/v), Chloride Free, Indicator for Argentometric Titrations, Ricca Chemical
$242.63 - $438.25
Chemical Identifiers
| CAS | 7789-00-6 |
|---|---|
| Molecular Formula | CrK2O4 |
| Molecular Weight (g/mol) | Mixture |
| MDL Number | MFCD00011368 |
| InChI Key | XMXNVYPJWBTAHN-UHFFFAOYSA-N |
| Synonym | potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687 |
| PubChem CID | 24597 |
| ChEBI | CHEBI:75249 |
| IUPAC Name | dipotassium dioxochromiumbis(olate) |
| SMILES | [K+].[K+].[O-][Cr]([O-])(=O)=O |
Chemical Identifiers
| 7789-00-6 | |
| Mixture | |
| XMXNVYPJWBTAHN-UHFFFAOYSA-N | |
| 24597 | |
| dipotassium dioxochromiumbis(olate) |
| CrK2O4 | |
| MFCD00011368 | |
| potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687 | |
| CHEBI:75249 | |
| [K+].[K+].[O-][Cr]([O-])(=O)=O |
Specifications
| 7789-00-6 , 7732-18-5 , 7761-88-8 | |
| 9.27,93.0,7.0E-4 | |
| CrK2O4 | |
| potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687 | |
| [K+].[K+].[O-][Cr]([O-])(=O)=O | |
| Mixture | |
| CHEBI:75249 | |
| Natural Poly Bottle | |
| Yellow | |
| Liquid |
| 8.91,89.0,7.0E-4 | |
| 10% (w/v) | |
| MFCD00011368 | |
| XMXNVYPJWBTAHN-UHFFFAOYSA-N | |
| dipotassium dioxochromiumbis(olate) | |
| 24597 | |
| Indicator | |
| Aqueous Solution | |
| 500 mL | |
| Potassium Chromate, Indicator for Argentometric Titrations, Test Solution |