missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chromate TS, 10% (w/v), Chloride Free, Indicator for Argentometric Titrations, Ricca Chemical

Catalog No. 600532
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
600532 1 L Natural Poly Bottle
600516 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 600532 Supplier Ricca Chemical Company Supplier No. 600532
Only null left

Chemical Identifiers

CAS 7789-00-6 , 7732-18-5 , 7761-88-8
Molecular Formula CrK2O4
Molecular Weight (g/mol) Mixture
MDL Number MFCD00011368
InChI Key XMXNVYPJWBTAHN-UHFFFAOYSA-N
Synonym potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687
PubChem CID 24597
ChEBI CHEBI:75249
IUPAC Name dipotassium dioxochromiumbis(olate)
SMILES [K+].[K+].[O-][Cr]([O-])(=O)=O

Specifications

CAS Min % 8.91, 89.0, 7.0E-4
CAS Max % 9.27, 93.0, 7.0E-4
Concentration 10% (w/v)
Grade Indicator
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Color Yellow
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Potassium Chromate, Indicator for Argentometric Titrations, Test Solution