missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chromate Indicator, For Chloride, Certified, 5% (w/v), LabChem™

$75.11 - $346.79

Chemical Identifiers

CAS 7732-18-5
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
LC188501
View Documents
LabChem
LC188501
500 mL Poly Bottle
Each for $75.11
Only null left
 
LC188502
View Documents
LabChem
LC188502
1 L Poly Bottle
Each for $117.14
Only null left
 
LC188504
View Documents
LabChem
LC188504
4 L Poly Bottle
Each for $346.79
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5

Specifications

7732-18-5 , 7789-00-6 , 7761-88-8
K2CrO4
Passes Test
5% (w/v)
Poly Bottle
Passes Test
1.05g/mL
500 mL
Potassium Chromate Indicator, For Chloride
94.9992,5,0.0008
Soluble in water
194.2
Certified
1.05g/mL
Contains hexavalent Chromium
Orange/Yellow
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
May cause genetic defects.
May cause cancer.
May cause damage to organs (blood, kidneys, liver) through prolonged or repeated exposure.
Very toxic to aquatic life.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist.
Wear protective gloves, protective clothing, eye protection, face protection.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If on skin: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
Take off contaminated clothing and wash it before reuse.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

Recommended Storage : Room Temperature

SDS
Documents

Documents