missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chromate Indicator, For Chloride, Certified, 5% (w/v), LabChem™

Supplier:  LabChem LC188504

 View more versions of this product

Catalog No. LC188504

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Chromate Indicator
7732-18-5 , 7789-00-6 , 7761-88-8
K2CrO4
Passes Test
5% (w/v)
Poly Bottle
Passes Test
1.05g/mL
4 L
For Chloride
94.9992,5,0.0008
Soluble in water
194.2
Certified
1.05g/mL
Contains hexavalent Chromium
Orange/Yellow
Liquid

Chemical Identifiers

7732-18-5
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
May cause genetic defects.
May cause cancer.
May cause damage to organs (blood, kidneys, liver) through prolonged or repeated exposure.
Very toxic to aquatic life.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist.
Wear protective gloves, protective clothing, eye protection, face protection.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If on skin: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
Take off contaminated clothing and wash it before reuse.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions