missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chromate, For Silica, 0.063%, LabChem™

Catalog No. LC188401
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC188401 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC188401 Supplier LabChem Supplier No. LC188401
Only null left

Chemical Identifiers

CAS 7789-00-6 , 7732-18-5
Molecular Formula CrK2O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00011368
InChI Key XMXNVYPJWBTAHN-UHFFFAOYSA-N
Synonym potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687
PubChem CID 24597
ChEBI CHEBI:75249
IUPAC Name dipotassium dioxochromiumbis(olate)
SMILES [K+].[K+].[O-][Cr]([O-])(=O)=O

Specifications

Color Yellow
CAS Max % 99.94, 0.06
Linear Formula K2CrO4
Density 1g/mL
Solubility Information Soluble in water
Formula Weight 194.2
Percent Purity 0.063%
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Decomposition Information Contains hexavalent Chromium
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Potassium Chromate, For Silica
Show More Show Less
Hazard Statement GHS H Statement
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature