missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chromate, ACS Reagent Grade, Ricca Chemical

Catalog No. RDCP0390100
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCP0390100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCP0390100 Supplier Ricca Chemical Company Supplier No. RDCP0390100B1
Only null left

Chemical Identifiers

CAS 7789-00-6
Molecular Formula CrK2O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00011368
InChI Key XMXNVYPJWBTAHN-UHFFFAOYSA-N
Synonym potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687
PubChem CID 24597
ChEBI CHEBI:75249
IUPAC Name dipotassium dioxochromiumbis(olate)
SMILES [K+].[K+].[O-][Cr]([O-])(=O)=O

Specifications

Color Yellow
CAS Min % 98.0
CAS Max % 100.0
Physical Form Crystals or Powder
Quantity 100 g
Percent Purity ≥ 99%
Grade ACS
Packaging White Poly Wide Mouth Bottle
Chemical Name or Material Potassium Chromate
ShelfLife 60 months