missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chromate, 5% (w/v), Chloride Free, Indicator for Argentometric Titrations, Ricca Chemical
Specifications
Specifications
| Color | Yellow |
| CAS Min % | 4.67, 93.0, 7.0E-4 |
| CAS Max % | 4.86, 97.0, 7.0E-4 |
| Percent Purity | 5% |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| Solvent or Matrix | Aqueous Solution |
| pH | Alkaline |
| Quantity | 1 L |
| Physical Form | Liquid |
| Show More |