missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Saturated, Electrode Filling Solution, Certified, ≥ 4.0M, LabChem™

Catalog No. p-5388493
Change view
Click to view available options
Quantity:
125 mL
500 mL
1 L
10 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC188007 125 mL
LC188001 500 mL
LC188002 1 L
LC188006 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. LC188007 Supplier LabChem Supplier No. LC188007
Only null left

Chemical Identifiers

CAS 7447-40-7 , 7732-18-5
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Color Colorless
CAS Max % 73.6, 26.4
Linear Formula KCl
Concentration ≥ 4.0M
Density 1.18g/mL
Solubility Information Soluble in water
Formula Weight 74.55
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.18g/mL
Decomposition Information Hydrogen chloride; Potassium oxide
Quantity 125 mL
Physical Form Liquid
Chemical Name or Material Potassium Chloride Saturated, Electrode Filling Solution
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature