missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Saturated, Electrode Filling Solution, Certified, ≥ 4.0M, LabChem™

Supplier:  LabChem LC188002

 View more versions of this product

Catalog No. LC188002


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Chloride Saturated
Electrode Filling Solution
73.6,26.4
KCl
≥ 4.0M
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride
4873
74.55
Passes Test
1.18g/mL
1 L
Colorless
7447-40-7,7732-18-5
ClK
MFCD00011360
1.18g/mL
Soluble in water
[Cl-].[K+]
74.55
CHEBI:32588
Certified
Poly Bottle
Hydrogen chloride; Potassium oxide
Liquid

Chemical Identifiers

7447-40-7
74.55
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
4873
potassium chloride
ClK
MFCD00011360
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
CHEBI:32588
[Cl-].[K+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions