missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride Saturated, Electrode Filling Solution, Certified, ≥ 4.0M, LabChem™
Supplier: LabChem LC188001
Specifications
| Potassium Chloride Saturated | |
| Electrode Filling Solution | |
| 73.6,26.4 | |
| KCl | |
| ≥ 4.0M | |
| potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| potassium chloride | |
| 4873 | |
| 74.55 | |
| Passes Test | |
| 1.18g/mL | |
| Hydrogen chloride; Potassium oxide | |
| Liquid |
| Colorless | |
| 7447-40-7,7732-18-5 | |
| ClK | |
| MFCD00011360 | |
| 1.18g/mL | |
| Soluble in water | |
| [Cl-].[K+] | |
| 74.55 | |
| CHEBI:32588 | |
| Certified | |
| Poly Bottle | |
| 0.895cSt | |
| 500 mL |
Chemical Identifiers
| 7447-40-7 | |
| 74.55 | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| 4873 | |
| potassium chloride |
| ClK | |
| MFCD00011360 | |
| potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
| CHEBI:32588 | |
| [Cl-].[K+] |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature