missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Reference Solution D, 146.93 μS/cm at 25°C, 0.001 M, Ricca Chemical

Catalog No. 589616
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
589616 500 mL Natural Poly Bottle
589632 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 589616 Supplier Ricca Chemical Company Supplier No. 589616
Only null left

Chemical Identifiers

CAS 7447-40-7 , 7732-18-5
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 0.001Molar
CAS Min % 98.0, 0.01
CAS Max % 100.0, 0.01
Physical Form Liquid
Color Colorless
pH Approximately 7
Boiling Point 100°C
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 500 mL
Chemical Name or Material Potassium Chloride Reference Solution D, 146.93μS/cm (146.93μmho/cm) at 25°C
Show More Show Less