missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Reference Solution C, 1408.8 μS/cm at 25°C, 0.01 M, Ricca Chemical

Catalog No. 589532
Change view
Click to view available options
Quantity:
500 mL
1 L
10 L
Packaging:
Cubitainer
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
589532 1 L Natural Poly Bottle
589516 500 mL Natural Poly Bottle
589525 10 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 589532 Supplier Ricca Chemical Company Supplier No. 589532

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7447-40-7 , 7732-18-5
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Packaging Natural Poly Bottle
CAS Min % 98.0, 0.07
CAS Max % 100.0, 0.07
Physical Form Liquid
Color Colorless
pH Approximately 7
Boiling Point 100°C
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 1 L
Concentration or Composition (by Analyte or Components) 0.01M
Chemical Name or Material Potassium Chloride Reference Solution C, 1408.8μS/cm (1408.8μmho/cm) at 25°C
Show More Show Less