missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride, Granular, USP, 99-100.5%, Spectrum™ Chemical
PO170, 7447-40-7, KCl
$1287.17 - $6079.90
Chemical Identifiers
| CAS | 7447-40-7 |
|---|---|
| Molecular Formula | ClK |
| Molecular Weight (g/mol) | 74.55 |
| MDL Number | MFCD00011360 |
| InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
| IUPAC Name | potassium chloride |
| SMILES | [Cl-].[K+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18605518
|
Spectrum Chemical Mfg Cor
PO17025KG |
2.5 kg | Poly Bottle |
Each for $1,287.17
|
|
||||
|
18605517
|
Spectrum Chemical Mfg Cor
PO170500GMCS6 |
6 x 500 g | Poly Bottle |
Case of 6 Each for $2,137.63
|
|
||||
|
18605519
|
Spectrum Chemical Mfg Cor
PO17025KGCS4 |
4 x 2.5 kg | Poly Bottle |
Case of 4 Each for $3,879.10
|
|
||||
|
18605520
|
Spectrum Chemical Mfg Cor
PO17012KG |
12 kg | Poly Pail |
Each for $2,072.17
|
|
||||
|
18605521
|
Spectrum Chemical Mfg Cor
PO17045KGBL |
45 kg | Fiber Drum |
Each for $3,858.00
|
|
||||
|
18605522
|
Spectrum Chemical Mfg Cor
PO1701135KGBL |
113.5 kg | Fiber Drum |
Each for $6,079.90
|
|
||||