missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride, Granular, USP, 99-100.5%, Spectrum™ Chemical
Description
Spectrum™ Chemical Potassium Chloride, Granular, USP is used to treat hypokalemia. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 2.5 kg |
| Percent Purity | 99 to 100.5% |
| Grade | USP |
| Loss on Drying | 0.01 |
| Packaging | Poly Bottle |