missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride, Granular, USP, 99-100.5%, Spectrum™ Chemical

PO170, 7447-40-7, KCl

Supplier:  Spectrum Chemical Mfg Cor PO17025KGCS4

 View more versions of this product

Catalog No. 18605519


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Potassium Chloride, Granular, USP is used to treat hypokalemia. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

7447-40-7
4 x 2.5 kg
MFCD00011360
[Cl-].[K+]
74.55
0.01
1
ClK
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride
99 to 100.5%
Poly Bottle

Chemical Identifiers

7447-40-7
74.55
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
[Cl-].[K+]
ClK
MFCD00011360
potassium chloride
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions