missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride, Electrode Filling Solution, Saturated with AgCl, Certified, 4.0M ±0.1M (4N), LabChem™
Supplier: LabChem LC140007
Specifications
| Potassium Chloride | |
| Electrode Filling Solution, Saturated with AgCl | |
| 25.65,74.3498,0.0002 | |
| KCl | |
| 4.0M ±0.1M (4N) | |
| potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| potassium chloride | |
| 4873 | |
| 74.55 | |
| Passes Test | |
| 1.18g/mL | |
| Hydrogen chloride; Potassium oxide | |
| Liquid |
| Colorless | |
| 7447-40-7 , 7732-18-5 , 7783-90-6 | |
| ClK | |
| MFCD00011360 | |
| 1.18g/mL | |
| Soluble in water | |
| [Cl-].[K+] | |
| 74.55 | |
| CHEBI:32588 | |
| Certified | |
| Poly Bottle | |
| Traceable to NIST | |
| 125 mL |
Chemical Identifiers
| 7447-40-7 | |
| 74.55 | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| 4873 | |
| potassium chloride |
| ClK | |
| MFCD00011360 | |
| potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
| CHEBI:32588 | |
| [Cl-].[K+] |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature