missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride Conductivity Standard, 600 μS/cm at 25°C, Ricca Chemical
$65.37 - $391.33
Chemical Identifiers
| CAS | 7447-40-7 |
|---|---|
| Molecular Formula | ClK |
| Molecular Weight (g/mol) | 74.55 |
| MDL Number | MFCD00011360 |
| InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
| PubChem CID | 4873 |
| ChEBI | CHEBI:32588 |
| IUPAC Name | potassium chloride |
| SMILES | [Cl-].[K+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
R5887600500
|
Ricca Chemical Company
R5887600500A |
500 mL |
Each for $65.37
|
|
|||||
|
R58876001A
|
Ricca Chemical Company
R58876001A |
1 L |
Each for $391.33
|
|
|||||
Chemical Identifiers
| 7447-40-7 | |
| 74.55 | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| CHEBI:32588 | |
| [Cl-].[K+] |
| ClK | |
| MFCD00011360 | |
| 4873 | |
| potassium chloride |
Specifications
| Natural Poly Bottle | |
| 99.96,0.04 | |
| 1 | |
| ClK | |
| Colorless | |
| 0°C | |
| Miscible with water | |
| Conductivity Standard | |
| [Cl-].[K+] | |
| 74.55 | |
| 4873 | |
| Potassium Chloride Conductivity Standard, 600μS/cm at 25°C |
Safety and Handling
ShelfLife : 18 months
Recommended Storage : Ambient